Skip to Content
Merck
All Photos(1)

Documents

778362

Sigma-Aldrich

5-Methyl-5-allyloxycarbonyl-1,3-dioxan-2-one

97%

Synonym(s):

5-Methyl-2-oxo-1,3-dioxane-5-carboxylic acid, 2-propen-1-yl ester, Allyl 5-methyl-2-oxo-1,3-dioxane-5-carboxylate, Allyl functionalized carbonate monomer, MAC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12O5
CAS Number:
Molecular Weight:
200.19
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

65-70 °C

storage temp.

−20°C

SMILES string

O=C1OCC(C(OCC=C)=O)(C)CO1

InChI

1S/C9H12O5/c1-3-4-12-7(10)9(2)5-13-8(11)14-6-9/h3H,1,4-6H2,2H3

InChI key

OPWZLMZGHGAFNG-UHFFFAOYSA-N

Application

This monomer contains a pendant allyl group for thiol-ene click chemistry; to be copolymerized with lactides or glycolides for biodegradable polymers containing pendant functional groups.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kerry A Hamilton et al.
Water research, 119, 288-303 (2017-05-14)
A quantitative microbial risk assessment (QMRA) of opportunistic pathogens Legionella pneumophila (LP) and Mycobacterium avium complex (MAC) was undertaken for various uses of roof-harvested rainwater (RHRW) reported in Queensland, Australia to identify appropriate usages and guide risk management practices. Risks
Nerve Zhou et al.
PloS one, 12(3), e0173318-e0173318 (2017-03-11)
The Crabtree positive yeasts, such as Saccharomyces cerevisiae, prefer fermentation to respiration, even under fully aerobic conditions. The selective pressures that drove the evolution of this trait remain controversial because of the low ATP yield of fermentation compared to respiration.
D J Eleveld et al.
British journal of anaesthesia, 120(5), 942-959 (2018-04-18)
Pharmacokinetic (PK) and pharmacodynamic (PD) models are used in target-controlled-infusion (TCI) systems to determine the optimal drug administration to achieve a desired target concentration in a central or effect-site compartment. Our aim was to develop a PK-PD model for propofol
Kanwal Ashraf et al.
Molecular biology and evolution, 36(5), 1037-1055 (2019-02-24)
Epigenetic information, which can be passed on independently of the DNA sequence, is stored in part in the form of histone posttranslational modifications and specific histone variants. Although complexes necessary for deposition have been identified for canonical and variant histones
M Coburn et al.
British journal of anaesthesia, 120(1), 127-137 (2018-02-06)
Postoperative delirium occurs frequently in elderly hip fracture surgery patients and is associated with poorer overall outcomes. Because xenon anaesthesia has neuroprotective properties, we evaluated its effect on the incidence of delirium and other outcomes after hip fracture surgery. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service