Skip to Content
Merck
All Photos(1)

Documents

774782

Sigma-Aldrich

Methyl 5-bromo-2-methoxypyridine-3-carboxylate

97%

Synonym(s):

5-Bromo-2-methoxypyridine-3-carboxylic acid methyl ester, Methyl 5-bromo-2-methoxynicotinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8BrNO3
CAS Number:
Molecular Weight:
246.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

100-102 °C

storage temp.

2-8°C

SMILES string

BrC1=CC(C(OC)=O)=C(OC)N=C1

InChI

1S/C8H8BrNO3/c1-12-7-6(8(11)13-2)3-5(9)4-10-7/h3-4H,1-2H3

InChI key

OLQOVEOEHQDKSC-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service