Skip to Content
Merck
All Photos(2)

Key Documents

772356

Sigma-Aldrich

Lactose octaacetate

95%

Synonym(s):

4-O-(2,3,4,6-Tetra-O-acetyl-β-D-galactopyranosyl)-1,2,3,6-tetra-O-acetyl-D-glucopyranose

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€94.20
100 G
€571.00

€94.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€94.20
100 G
€571.00

About This Item

Empirical Formula (Hill Notation):
C28H38O19
CAS Number:
Molecular Weight:
678.59
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

€94.20


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

solid

mp

75-78 °C

storage temp.

2-8°C

SMILES string

[R]OC1C(O[R])[C@@H](O[R])C(CO[R])O[C@H]1O[C@H]2C(O[R])[C@@H](O[R])[C@H](O[R])OC2CO[R].[*]C(C)=O

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service