Skip to Content
Merck
All Photos(3)

Key Documents

772003

Sigma-Aldrich

Fmoc-D-Asn(Trt)-OH

97%, for peptide synthesis

Synonym(s):

Fmoc-N-trityl-D-asparagine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€49.80
5 G
€117.00
25 G
€324.00

€49.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€49.80
5 G
€117.00
25 G
€324.00

About This Item

Empirical Formula (Hill Notation):
C38H32N2O5
CAS Number:
Molecular Weight:
596.67
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

€49.80


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Fmoc-D-Asn(Trt)-OH, 97%

Quality Level

Assay

97%

form

powder or crystals

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

mp

211-216 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)[C@@H](CC(=O)NC(c1ccccc1)(c2ccccc2)c3ccccc3)NC(=O)OCC4c5ccccc5-c6ccccc46

InChI

1S/C38H32N2O5/c41-35(40-38(26-14-4-1-5-15-26,27-16-6-2-7-17-27)28-18-8-3-9-19-28)24-34(36(42)43)39-37(44)45-25-33-31-22-12-10-20-29(31)30-21-11-13-23-32(30)33/h1-23,33-34H,24-25H2,(H,39,44)(H,40,41)(H,42,43)/t34-/m1/s1

InChI key

KJYAFJQCGPUXJY-UUWRZZSWSA-N

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service