Skip to Content
Merck
All Photos(1)

Documents

768103

Sigma-Aldrich

Dichlorobis(di-tert-butylphenylphosphine)palladium(II)

95%

Synonym(s):

(SP-4-1)-Bis[bis(1,1-dimethylethyl)phenylphosphine]dichloropalladium, trans-Dichlorobis(di-tert-butylphenylphosphine)palladium, trans-Dichlorobis(di-tert-butylphenylphosphine)palladium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H46Cl2P2Pd
CAS Number:
Molecular Weight:
621.94
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

260-264 °C (decomposition)

SMILES string

Cl[Pd]Cl.CC(C)(C)P(c1ccccc1)C(C)(C)C.CC(C)(C)P(c2ccccc2)C(C)(C)C

InChI

1S/2C14H23P.2ClH.Pd/c2*1-13(2,3)15(14(4,5)6)12-10-8-7-9-11-12;;;/h2*7-11H,1-6H3;2*1H;/q;;;;+2/p-2

InChI key

QNNRAWADYXIGHE-UHFFFAOYSA-L

Application

Effective catalyst for cross-coupling reactions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xavier Guinchard et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(23), 5793-5798 (2009-04-28)
We describe the preparation of a series of new potassium trifluoroborates 1 and the study of their behaviour in a Pd(0)-catalyzed cross-coupling reaction. We found that compounds 1 are endowed with original properties as they behave as nucleophilic cross-coupling partners
Anil S Guram et al.
The Journal of organic chemistry, 72(14), 5104-5112 (2007-06-07)
The new air-stable PdCl2[PR2(Ph-R')]2 complexes, readily prepared from commercial reagents, exhibit unique efficiency as catalysts for the Suzuki-Miyaura coupling reactions of a variety of heteroatom-substituted heteroaryl chlorides with a diverse range of aryl/heteroaryl boronic acids. The coupling reactions catalyzed by
Anil S Guram et al.
Organic letters, 8(9), 1787-1789 (2006-04-21)
[reaction: see text] New air-stable PdCl(2){P(t)Bu(2)(p-R-Ph)}(2) (R = H, NMe(2), CF(3),) complexes represent simple, general, and efficient catalysts for the Suzuki-Miyaura cross-coupling reactions of aryl halides including five-membered heteroaryl halides and heteroatom-substituted six-membered heteroaryl chlorides with a diverse range of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service