Skip to Content
Merck
All Photos(1)

Documents

768081

Sigma-Aldrich

2-(Diphenylphosphino)-N,N,N-trimethylbenzylammonium triflate

96%

Synonym(s):

1-(2-(Diphenylphosphino)phenyl)-N,N,N-trimethylmethanaminium trifluoromethanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H25F3NO3PS
CAS Number:
Molecular Weight:
483.48
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

154-159 °C

functional group

phosphine

SMILES string

C[N+](C)(C)CC1=C(C=CC=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[O-]S(=O)(C(F)(F)F)=O

InChI

1S/C22H25NP.CHF3O3S/c1-23(2,3)18-19-12-10-11-17-22(19)24(20-13-6-4-7-14-20)21-15-8-5-9-16-21;2-1(3,4)8(5,6)7/h4-17H,18H2,1-3H3;(H,5,6,7)/q+1;/p-1

InChI key

OTAHBMHXZGOAGA-UHFFFAOYSA-M

Application

Phosphine ligand for Pd catalysis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service