Skip to Content
Merck
All Photos(1)

Key Documents

767824

Sigma-Aldrich

iPE-Quick Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€541.00

€541.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 KIT
€541.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.12

€541.00


Please contact Customer Service for Availability

Request more information

technique(s)

bio NMR: suitable
protein expression: suitable

Quality Level

shipped in

dry ice

storage temp.

−70°C

Related Categories

Application

iPE-Quick Kit is intended for the advanced confirmation of target protein expression utilizing E. Coli extract before the use of the iPE kit (Prod. No. 905089).

Packaging

For information on pricing and availability, please contact Stable Isotopes Customer Service.

Kit Components Also Available Separately

Product No.
Description
SDS

  • Control DNA (pUC-CAT)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service