Skip to Content
Merck
All Photos(1)

Key Documents

766992

Sigma-Aldrich

Diethyl 4-fluorobenzylphosphonate

97%

Synonym(s):

Diethyl (p-fluorophenyl)methanephosphonate, [(4-Fluorophenyl)methyl]phosphonic acid diethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€31.20

€31.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€31.20

About This Item

Empirical Formula (Hill Notation):
C11H16FO3P
CAS Number:
Molecular Weight:
246.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€31.20


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

liquid

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.483

density

1.143 g/mL at 25 °C

functional group

fluoro
phosphonate

SMILES string

CCOP(=O)(Cc1ccc(F)cc1)OCC

InChI

1S/C11H16FO3P/c1-3-14-16(13,15-4-2)9-10-5-7-11(12)8-6-10/h5-8H,3-4,9H2,1-2H3

InChI key

FIYRZOAUPPNGAO-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service