Skip to Content
Merck
All Photos(1)

Key Documents

765007

Sigma-Aldrich

2,2′,7,7′-Tetrakis(N,N-diphenylamino)-9,9-spirobifluorene

99% (HPLC)

Synonym(s):

N2,N2,N2′,N2′,N7,N7,N7′,N7′-octaphenyl-9,9′-spirobi[9H-fluorene]-2,2′,7,7′-tetramine, Spiro-TAD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C73H52N4
CAS Number:
Molecular Weight:
985.22
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

99% (HPLC)

form

powder

mp

275-280 °C

λmax

378 μm in THF

SMILES string

c1ccc(cc1)N(c2ccccc2)c3ccc4-c5ccc(cc5C6(c4c3)c7cc(ccc7-c8ccc(cc68)N(c9ccccc9)c%10ccccc%10)N(c%11ccccc%11)c%12ccccc%12)N(c%13ccccc%13)c%14ccccc%14

InChI

1S/C73H52N4/c1-9-25-53(26-10-1)74(54-27-11-2-12-28-54)61-41-45-65-66-46-42-62(75(55-29-13-3-14-30-55)56-31-15-4-16-32-56)50-70(66)73(69(65)49-61)71-51-63(76(57-33-17-5-18-34-57)58-35-19-6-20-36-58)43-47-67(71)68-48-44-64(52-72(68)73)77(59-37-21-7-22-38-59)60-39-23-8-24-40-60/h1-52H

InChI key

MQRCTQVBZYBPQE-UHFFFAOYSA-N

Application

Hole transport materials comprised of a spiro center between two charge transport material (CTM) moieties. This molecular conformation improves the thermal stability of the amorphous state, without significantly altering the charge-transport properties[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Advanced Materials, 12, 1060-1063 (2000)
Michael Grimann et al.
The journal of physical chemistry. B, 124(26), 5507-5516 (2020-06-05)
Understanding phase separation phenomena in blends of organic electron acceptor and donor materials is of special interest in the context of organic optoelectronic applications. In this study, we focus on the phase behavior of a special class of spiro-linked compounds

Articles

While dye sensitization as the basis for color photography has been accepted for a very long time,1 attempts to use this principle for the conversion of solar light to electricity generally had resulted only in very low photocurrents, below 100 nA/cm

For several decades, the need for an environmentally sustainable and commercially viable source of energy has driven extensive research aimed at achieving high efficiency power generation systems that can be manufactured at low cost.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service