Skip to Content
Merck
All Photos(2)

Key Documents

764787

Sigma-Aldrich

Poly(lactide-co-glycolide)-block-poly(ethylene glycol)-block-poly(lactide-co-glycolide)

average Mn (1,000-1,000-1,000), lactide:glycolide 50:50

Synonym(s):

PLGA-PEG-PLGA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

description

typical PEG PDI<1.1; overall PDI<1.2 (THF, PEO)

form

liquid

feed ratio

lactide:glycolide 50:50

mol wt

PEG average Mn 1,000
PLGA average Mn 2000
average Mn (1,000-1,000-1,000)

degradation timeframe

1-2 weeks

PDI

<1.2

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

PLGA-PEG-PLGA is an amphiphilic triblock copolymer which can self-assemble into micelles in aqueous medium due to the hydrophobic interactions present in the hydrophobic segments. The PEG segment imparts hydrophilicity and improves the biocompatibility of the copolymer. The PLGA segment forms a hydrophobic core and can solubilize hydrophobic drugs. These copolymers are widely used as nanocarriers for the sustained release of drugs.

Application

Used in the synthesis of targeted nanoparticles which are used for differential delivery and controlled release of drugs.

Features and Benefits

Biocompatible, degradable, thermosensitive, high stability, small size (<200 nm) and properties can be easily modified.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhimei Song et al.
Journal of colloid and interface science, 354(1), 116-123 (2010-11-04)
The aim of this study was to assess the potential of new copolymeric micelles to modify the pharmacokenetics and tissue distribution of Curcumin (CUR), a hydrophobic drug. In the present study, a poly (d,l-lactide-co-glycolide)-b-poly(ethylene glycol)-b-poly(d,l-lactide-co-glycolide) (PLGA-PEG-PLGA) copolymer was synthesized and
Frank Gu et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(7), 2586-2591 (2008-02-15)
There has been progressively heightened interest in the development of targeted nanoparticles (NPs) for differential delivery and controlled release of drugs. Despite nearly three decades of research, approaches to reproducibly formulate targeted NPs with the optimal biophysicochemical properties have remained
PLGA-PEG Encapsulated sitamaquine nanoparticles drug delivery system against Leishmania donovani
Kumara, R., Sahoo, G. C., Pandeya, K., Dasa, V. N. R., Yousuf, M., Ansaria, S. R., &amp; Dasa, P.
Journal of Scientific and Innovative Research, 3(1), 85-90 (2014)

Articles

Local delivery of bioactive molecules using an implantable device can decrease the amount of drug dose required as well as non-target site toxicities compared to oral or systemic drug administration.

AliAliphatic polyesters, including polylactide and polyglycolide, are biodegradable polymers widely used in medical applications.

Humankind has utilized protein materials throughout its existence, starting with the use of materials such as wool and silk for warmth and protection from the elements and continuing with the use of recombinant DNA techniques to synthesize proteins with unique and useful properties.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service