Skip to Content
Merck
All Photos(1)

Key Documents

762865

Sigma-Aldrich

2-(Diisopropylphosphino)ethylamine solution

10 wt. % in THF

Synonym(s):

2-(Diisopropylphosphino)ethylamine, 2-[Bis(1-methylethyl)phosphino]ethanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H20NP
CAS Number:
Molecular Weight:
161.22
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

form

liquid

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

concentration

10 wt. % in THF

refractive index

n20/D 1.416
n20/D 1.489

density

0.872 g/mL at 25 °C
0.879 g/mL at 25 °C

functional group

phosphine

SMILES string

CC(C)P(CCN)C(C)C

InChI

1S/C8H20NP/c1-7(2)10(6-5-9)8(3)4/h7-8H,5-6,9H2,1-4H3

InChI key

QUVUVMRUSAKOPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for synthesis of ruthenium hydrogenation catalysts

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

3.2 °F

Flash Point(C)

-16 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service