Skip to Content
Merck
All Photos(1)

Key Documents

761419

Sigma-Aldrich

(R)-(4,4′,6,6′-Tetramethoxybiphenyl-2,2′-diyl)bis(bis(3,5-di-tert-butyl-4-methoxyphenyl)phosphine

97%

Synonym(s):

(R)-DTBM-Garphos

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€124.00
500 MG
€491.00

€124.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€124.00
500 MG
€491.00

About This Item

Empirical Formula (Hill Notation):
C76H108O8P2
CAS Number:
Molecular Weight:
1211.61
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

€124.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

solid

optical activity

[α]22/D +18.0°, c = 0.5% in chloroform

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

functional group

phosphine

SMILES string

COc1cc(OC)c(c(c1)P(c2cc(c(OC)c(c2)C(C)(C)C)C(C)(C)C)c3cc(c(OC)c(c3)C(C)(C)C)C(C)(C)C)-c4c(OC)cc(OC)cc4P(c5cc(c(OC)c(c5)C(C)(C)C)C(C)(C)C)c6cc(c(OC)c(c6)C(C)(C)C)C(C)(C)C

InChI

1S/C76H108O8P2/c1-69(2,3)51-37-47(38-52(65(51)81-29)70(4,5)6)85(48-39-53(71(7,8)9)66(82-30)54(40-48)72(10,11)12)61-35-45(77-25)33-59(79-27)63(61)64-60(80-28)34-46(78-26)36-62(64)86(49-41-55(73(13,14)15)67(83-31)56(42-49)74(16,17)18)50-43-57(75(19,20)21)68(84-32)58(44-50)76(22,23)24/h33-44H,1-32H3

InChI key

KACYLFSRRUJDSY-UHFFFAOYSA-N

Related Categories

Application

Versatile chiral diphosphine ligand.

Legal Information

Sold under license from Kanata Chemical Technologies Inc. for research purposes only.
Garphos is a trademark of Kanata Chemical Technologies, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service