Skip to Content
Merck
All Photos(1)

Key Documents

760943

Sigma-Aldrich

Poly(ethylene glycol), 8 acetylene dendron, generation 2

average Mn 21,000

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

description

acetylene surface groups

form

solid

mol wt

PEG average Mn 20,000 (n~450; esterified with 8 acetylene groups)
Mn 16,000-21,000 (by NMR)
average Mn 21,000

feature

generation 2

no. Surface Groups

8

transition temp

Tm 56-60 °C

Mw/Mn

≤1.4

functional group

alkyne

Looking for similar products? Visit Product Comparison Guide

General description

Dendrimers and dendrons are symmetrically branched polymer structures that possess a well-defined spatial distribution of functional groups. Dendrimers are chemically modified by attaching poly ethylene glycol (PEG) molecule to increase its solubility and prolong circulation half-life. “Clickable groups”, such as primary acetylenes, are introduced to the mono dispersed dendritic structures, using “click” chemistry.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Applications of Orthogonal, ?click? Chemistries in the synthesis of functional soft materials
Chemical Reviews, 109 (11), 5620-5686 null
Huiyuan Li et al.
Molecules (Basel, Switzerland), 18(8), 9797-9817 (2013-08-21)
Click chemistry is an efficient and chemoselective synthetic method for coupling molecular fragments under mild reaction conditions. Since the advent in 2001 of methods to improve stereochemical conservation, the click chemistry approach has been broadly used to construct diverse chemotypes
Degradable Poly (ethylene glycol) hydrogels for 2 D and 3D cell culture
Andrea M Kasko
Material Matters, 6 (3), 67-75 null
Biocompatible Dendritic Building Blocks for Advanced Biomedical Reasearch
Michael Malkoch and Andreas M Nystrom
Material Matters, 7(3), 47-49 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service