Skip to Content
Merck
All Photos(1)

Documents

760250

Sigma-Aldrich

4H-Thieno[3,2-b]pyrrole-5-carboxylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5NO2S
Molecular Weight:
167.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

201-205 °C

SMILES string

OC(=O)c1cc2sccc2[nH]1

InChI

1S/C7H5NO2S/c9-7(10)5-3-6-4(8-5)1-2-11-6/h1-3,8H,(H,9,10)

InChI key

PMHDSACGRKBACK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Megumi Iwasaki et al.
Biomedical chromatography : BMC, 30(3), 384-389 (2015-07-16)
Elution profiles of kynurenic acid (KYNA) and 7-chlorokynurenic acid (Cl-KYNA) were examined by high-performance liquid chromatography (HPLC) using a triazole-bonded stationary phase column (Cosmosil® HILIC) under isocratic elution of a mobile phase consisting of CH3 CN-aqueous 10 mm ammonium formate between

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service