Skip to Content
Merck
All Photos(2)

Key Documents

757942

Sigma-Aldrich

AlCuFe quasicrystal

powder, 55-75 μm, ≥99.9% trace metals basis

Synonym(s):

Al-Cu-Fe, Icosahedral-AlCuFe, i-AlCuFe

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Al65Cu23Fe12
Molecular Weight:
3885.50
UNSPSC Code:
12352103
NACRES:
NA.23

Assay

≥99.9% trace metals basis

form

powder

particle size

55-75 μm

Looking for similar products? Visit Product Comparison Guide

Application

AlCuFe quasicrystalline powder with narrow size distribution is suitable for thermal spraying, plasma spraying, coatings and thin film deposition, besides showing unique proerties arising from its quasicrystalline nature.

Pictograms

FlameEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Salimon, A. I.; Korsunky, A. M.; Shelekhov, E. V.; et al
Acta Materialia, 49, 1821-1821 (2001)
Moskalewicz. T.; Kot, M.; Wendler, B
Applied Surface Science, 258, 848-848 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service