Skip to Content
Merck
All Photos(1)

Key Documents

757683

Sigma-Aldrich

Bis(trifluoromethanesulfonyl)methane

97%

Synonym(s):

Bis(trifluoromethylsulfonyl)methane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H2F6O4S2
CAS Number:
Molecular Weight:
280.17
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

32-36 °C

functional group

fluoro
sulfone

SMILES string

FC(F)(F)S(=O)(=O)CS(=O)(=O)C(F)(F)F

InChI

1S/C3H2F6O4S2/c4-2(5,6)14(10,11)1-15(12,13)3(7,8)9/h1H2

InChI key

QBOFWVRRMVGXIG-UHFFFAOYSA-N

Application

A useful, solid, non-hygroscopic, nonoxidizing, soluble in organic solvents ligand for Lanthanide chemistry

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service