Skip to Content
Merck
All Photos(1)

Key Documents

757594

Sigma-Aldrich

Poly(styrene)-block-poly(acrylic acid), azide terminated

average Mn 8,600, PS:PAA block ratio 2:1

Synonym(s):

PS-b-PAA, Polystyrene-polyacrylic acid polymersome forming polymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(C6H5)[CH2CH(C6H5)]x [CH2CH(CO2H)]y N3
UNSPSC Code:
12162002

form

powder

mol wt

Mn 2,600-2,950 (polyacrylic acid)
Mn 5,500-6,000 (polystyrene)
Mn 8,100-9,100
average Mn 8,600

mp

>300 °C

PDI

<1.2

Looking for similar products? Visit Product Comparison Guide

Application

Polymersome forming block copolymer with an azide end-group for click chemistry.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The development of drugs that target specific locations within the human body remains one of the greatest challenges in biomedicine today.

Professor Robert K. Prud’homme introduces flash nanoprecipitation (FNP) for nanoparticle fabrication, which is a scalable, rapid mixing process for nanoparticle formulations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service