Skip to Content
Merck
All Photos(1)

Key Documents

757241

Sigma-Aldrich

9-(9-Heptadecanyl)-9H-carbazole-2,7-diboronic acid bis(pinacol) ester

97%

Synonym(s):

9-(1-Octylnonyl)-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H65B2NO4
CAS Number:
Molecular Weight:
657.58
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

powder

SMILES string

CCCCCCCCC(CCCCCCCC)n1c2cc(ccc2c3ccc(cc13)B4OC(C)(C)C(C)(C)O4)B5OC(C)(C)C(C)(C)O5

InChI

1S/C41H65B2NO4/c1-11-13-15-17-19-21-23-33(24-22-20-18-16-14-12-2)44-36-29-31(42-45-38(3,4)39(5,6)46-42)25-27-34(36)35-28-26-32(30-37(35)44)43-47-40(7,8)41(9,10)48-43/h25-30,33H,11-24H2,1-10H3

InChI key

XMKFCPVHTTWWCK-UHFFFAOYSA-N

Application

9-(9-Heptadecanyl)-9H-carbazole-2,7-diboronic acid bis(pinacol) ester can be used in the synthesis of organic semiconducting polymers for optoelectronic applications such as organic photovoltaics.[1][2][3][4]
This material is used in the formation of low band gap semiconducting materials for low cost plastic solar cells

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multipurpose selenophene containing conjugated polymers for optoelectronic applications
Onk I, et al.
Journal of Macromolecular Science, Part A: Pure and Applied Chemistry, 54(3), 133-139 (2017)
Photoactive Donor-Acceptor Composite Nanoparticles Dispersed in Water
Parrenin L, et al.
Langmuir, 33(6), 1507-1515 (2017)
Low Bandgap Semiconducting Copolymer Nanoparticles by Suzuki Cross-Coupling Polymerization in Alcoholic Dispersed Media
Parrenin L, et al.
Macromolecular Rapid Communications, 36(20), 1816-1821 (2015)
Incorporation of different conjugated linkers into low band gap polymers based on 5, 6-Bis (octyloxy)-2, 1, 3 benzooxadiazole for tuning optoelectronic properties
Goker S, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 54(16), 2459-2467 (2016)

Articles

Bifidobacteria-selective media detects beneficial bacteria, aiding quality control in microbiology labs.

Cost-effective molecular biology method for lactobacilli detection is presented as an alternative to PCR-based methods.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service