Skip to Content
Merck
All Photos(1)

Key Documents

755826

Sigma-Aldrich

4-Acetoxyphenethyl acrylate

Synonym(s):

2-Propenoic acid, 2-[4-(acetoxy)phenyl]ethyl ester, EC-234, Phenyl ester epoxy curative hybrid, R1146

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14O4
CAS Number:
Molecular Weight:
234.25
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

liquid

refractive index

n20/D 1.511

density

1.131 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(=O)Oc1ccc(CCOC(=O)C=C)cc1

InChI

1S/C13H14O4/c1-3-13(15)16-9-8-11-4-6-12(7-5-11)17-10(2)14/h3-7H,1,8-9H2,2H3

InChI key

UPRJCBCFGSJZJU-UHFFFAOYSA-N

Application

  • Hybrid cure
  • Low viscosity
  • well-suited for UV Adhesives and B-stageable adhesives

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service