Skip to Content
Merck
All Photos(1)

Key Documents

753262

Sigma-Aldrich

5-(Trifluoromethyl)pyridine-2-carbonyl chloride

97%

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€103.00

€103.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€103.00

About This Item

Empirical Formula (Hill Notation):
C7H3ClF3NO
CAS Number:
Molecular Weight:
209.55
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€103.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.474

density

1.468 g/mL at 25 °C

functional group

acyl chloride
fluoro

SMILES string

FC(F)(F)c1ccc(nc1)C(Cl)=O

InChI

1S/C7H3ClF3NO/c8-6(13)5-2-1-4(3-12-5)7(9,10)11/h1-3H

InChI key

XGGHEBGOZWCTKF-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service