Skip to Content
Merck
All Photos(1)

Key Documents

753076

Bis(ethylcyclopentadienyl)cobalt(II)

packaged for use in deposition systems

Synonym(s):

Bis(ethylcyclopentadienyl)cobalt(II), (EtCp)2Co, 1,1′-Diethylcobaltocene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18Co
CAS Number:
Molecular Weight:
245.23
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

liquid

density

1.516 g/mL at 25 °C (lit.)

SMILES string

[Co].CC[C]1[CH][CH][CH][CH]1.CC[C]2[CH][CH][CH][CH]2

InChI

1S/2C7H9.Co/c2*1-2-7-5-3-4-6-7;/h2*3-6H,2H2,1H3;

InChI key

YHCQFTZSIGZRTR-UHFFFAOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hunde; E.T.; Watkins; J.J.
Chemistry of Materials, 16, 498-498 (2004)
Rushworth; S.
Material Matters, 5, 94-94 (2010)
Lain-Jong Li et al.
Nature materials, 4(6), 481-485 (2005-05-24)
Encapsulation of organic molecules in carbon nanotubes has opened a new route for the fabrication of hybrid nanostructures. Here we show that diameter-selective encapsulation of two metallocene compounds bis(cyclopentadienyl) cobalt and bis(ethylcyclopentadienyl) cobalt has been observed in single-walled carbon nanotubes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service