Skip to Content
Merck
All Photos(1)

Key Documents

751960

Sigma-Aldrich

Yttrium(III) chloride

vacuum purified, anhydrous, beads, -10 mesh, 99.99% trace metals basis

Synonym(s):

Yttrium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
YCl3
CAS Number:
Molecular Weight:
195.26
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Pricing and availability is not currently available.

grade

anhydrous

Assay

99.99% trace metals basis

form

beads

particle size

-10 mesh

mp

721 °C (lit.)

density

2.67 g/mL at 25 °C (lit.)

SMILES string

Cl[Y](Cl)Cl

InChI

1S/3ClH.Y/h3*1H;/q;;;+3/p-3

InChI key

PCMOZDDGXKIOLL-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Yttrium Chloride has been used a precursor for the synthesis of optically active crystals[1] and as a starting material for the synthesis of polymerization catalysts.[2]

Features and Benefits

Will react with potassium alkoxotitanates to form precursors to novel titanium-containing ceramics

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1B

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fanny Bonnet et al.
Inorganic chemistry, 44(24), 9046-9055 (2005-11-22)
Reaction of Na2O2NN' [H2O2NN' = (2-C5H4N)CH2N[2-HO-3,5-C6H2(t)Bu2]2] with M(BH4)3(THF)3 afforded the dimeric, rare-earth borohydride compounds [M(O2NN')(mu-BH4)(THF)n]2 [M = Y(III), n = 0.5 (1-Y); M = NdIII, n = 1 (1-Nd); M = SmIII, n = 0 (1-Sm)]. For comparison the chloride
Heer, S.; et al.
Advanced Materials, 16, 2102-2102 (2004)
S Hirano et al.
Toxicology and applied pharmacology, 121(2), 224-232 (1993-08-01)
Although radioactive yttrium (Y) has been used for medical treatment, little attention has been directed toward the toxicity of Y. We report time-course and dose-related changes in tissue distribution, subcellular localization, clearance, and acute toxicity of iv-injected yttrium chloride (YCl3)
Acceleration of the substitution of silanes with Grignard reagents by using either LiCl or YCl3/MeLi.
Naoki Hirone et al.
Angewandte Chemie (International ed. in English), 49(42), 7762-7764 (2010-09-08)
F Zhang et al.
Physical review letters, 101(14), 148101-148101 (2008-10-15)
Negatively charged globular proteins in solution undergo a condensation upon adding trivalent counterions between two critical concentrations C and C, C <C. This reentrant condensation behavior above C is caused by short-ranged electrostatic interactions between multivalent cations and acidic residues

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service