Skip to Content
Merck
All Photos(1)

Key Documents

749567

Sigma-Aldrich

4-Bromo-2,3-dimethylthiophene

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€25.80

€25.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€25.80

About This Item

Empirical Formula (Hill Notation):
C6H7BrS
CAS Number:
Molecular Weight:
191.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€25.80


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.571

density

1.495 g/mL at 25 °C

functional group

bromo

storage temp.

2-8°C

SMILES string

Cc1scc(Br)c1C

InChI

1S/C6H7BrS/c1-4-5(2)8-3-6(4)7/h3H,1-2H3

InChI key

GJHAWQJOXYVXFF-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

199.9 °F

Flash Point(C)

93.3 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service