Skip to Content
Merck
All Photos(3)

Key Documents

749133

Sigma-Aldrich

2-(Dodecylthiocarbonothioylthio)propionic acid

97%

Synonym(s):

2-(Dodecylthiocarbonothioylthio)propanoic acid, 2-{[(Dodecylsulfanyl)carbonothioyl]sulfanyl}propanoic acid, DoPAT

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€155.00
5 G
€501.00

€155.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€155.00
5 G
€501.00

About This Item

Empirical Formula (Hill Notation):
C16H30O2S3
CAS Number:
Molecular Weight:
350.60
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.23

€155.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

powder or crystals

mp

78-82 °C

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCSC(=S)SC(C)C(O)=O

InChI

1S/C16H30O2S3/c1-3-4-5-6-7-8-9-10-11-12-13-20-16(19)21-14(2)15(17)18/h14H,3-13H2,1-2H3,(H,17,18)

InChI key

CFCFZJHCTNKHGJ-UHFFFAOYSA-N

General description

Need help choosing the correct RAFT Agent? Please consult the RAFT Agent to Monomer compatibility table.

Application

RAFT agent for controlled radical polymerization; especially suited for the polymerization of styrene, acrylate and acrylamide monomers. Chain Transfer Agent (CTA)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Connor R Bilchak et al.
ACS nano (2020-11-21)
Polymer membranes are critical to many sustainability applications that require the size-based separation of gas mixtures. Despite their ubiquity, there is a continuing need to selectively affect the transport of different mixture components while enhancing mechanical strength and hindering aging.
Gordana Siljanovska Petreska et al.
Polymers, 12(6) (2020-06-04)
AB diblock waterborne copolymers made of styrene (St) and 2-ethylhexyl acrylate (2EHA) were synthesized by means of two-step reversible addition fragmentation chain transfer (RAFT) (mini)emulsion polymerization. Monofunctional asymmetric RAFT agent was used to initiate the polymerization. The hard polystyrene "A"
RAFT Agent Design and Synthesis
Keddie, D. J.; et al.
Macromolecules, 45, 5321-5342 (2012)
Massimo Benaglia et al.
Journal of the American Chemical Society, 131(20), 6914-6915 (2009-05-01)
The polymerization of most monomers that are polymerizable by radical polymerization can be controlled by the reversible addition-fragmentation chain transfer (RAFT) process. However, it is usually required that the RAFT agent be selected according to the types of monomer being

Articles

Low-cost, high-purity RAFT Agents are vital for industrial RAFT polymerization. We supply effective agents for this technology.

The modification of biomacromolecules, such as peptides and proteins, through the attachment of synthetic polymers has led to a new family of highly advanced biomaterials with enhanced properties.

Micro review of reversible addition/fragmentation chain transfer (RAFT) polymerization.

Protocols

We presents an article featuring procedures that describe polymerization of methyl methacrylate and vinyl acetate homopolymers and a block copolymer as performed by researchers at CSIRO.

We present an article about RAFT, or Reversible Addition/Fragmentation Chain Transfer, which is a form of living radical polymerization.

Polymerization via ATRP procedures demonstrated by Prof. Dave Haddleton's research group at the University of Warwick.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service