Skip to Content
Merck
All Photos(2)

Key Documents

749125

Sigma-Aldrich

Cobalt(II) oxo pivalate

greener alternative

Synonym(s):

Basic cobalt(II) pivalate, Cobalt, hexakis[μ-(2,2-dimethylpropanoato-κO:κO′)]hexakis[μ3-(2,2-dimethylpropanoato-κO:κO:κO′)]di-μ4-oxoocta-, Tetrameric Co cluster

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€515.00

€515.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€515.00

About This Item

Empirical Formula (Hill Notation):
C60H108Co8O26
CAS Number:
Molecular Weight:
1716.95
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

€515.00


Please contact Customer Service for Availability

Request a Bulk Order

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

SMILES string

CC(C)(C)C(=O)O[Co-]1O[Co-](O\C(=[O+]/[Co-](OC(=O)C(C)(C)C)O\C(=[O+]/[Co-](OC(=O)C(C)(C)C)OC(=O)C(C)(C)C)C(C)(C)C)C(C)(C)C)[O+]=C(O[Co-]2O[Co-](O\C(=[O+]\1)C(C)(C)C)\[O+]=C(\O[Co-](OC(=O)C(C)(C)C)\[O+]=C(\O[Co]OC(=[O+]2)C(C)(C)C)C(C)(C)C)C(C)(C)C)C(C)(C)C

InChI

1S/12C5H10O2.8Co.2O/c12*1-5(2,3)4(6)7;;;;;;;;;;/h12*1-3H3,(H,6,7);;;;;;;;;;/q;;;;;;;;;;;;4*+1;4*+2;;/p-12

InChI key

PBKALLJUIAKYSI-UHFFFAOYSA-B

General description

Cobalt(II) oxo pivalate is a dinuclear complex that is typically formed by the direct reaction of cobalt carbonate with pivalic acid, followed by crystallization from MeCN. Its structure has two Co(II) sites bridged by two 2,11-carboxylates (Harris notation) and an oxygen atom.[1]
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Synthesis of trinuclear complex [Co3(Piv)2L12L22] by reacting cobalt(II)pivalate with spin labeled Schiff base has been reported in a study.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic and structural studies of cobalt-pivalate complexes
Aromi G, et al.
Chemistry?A European Journal , 9(20), 5142-5161 (2003)
Redox-Induced Change in the Ligand Coordination Mode.
Ovcharenko V, et al.
Inorganic Chemistry, 53(19), 10033-10035 (2014)
Steffen Hausdorf et al.
Journal of the American Chemical Society, 132(32), 10978-10981 (2010-08-12)
A simple two-component procedure was developed to synthesize not only classical zinc-based IRMOFs represented by MOF-5 but also the cobalt and beryllium homologues of this most prominent MOF. The procedure is the first manifestation of mirroring the IRMOF series with
Steffen Hausdorf et al.
Dalton transactions (Cambridge, England : 2003), 7(7), 1107-1113 (2009-03-27)
An ethynylene diisophthalic acid linker molecule was synthesized and used to form a zinc carboxylate-based metal organic framework (MOF) with very large pores and unit cell volume resulting from the unusual combination of structurally different inorganic units forming the secondary
Steffen Hausdorf et al.
The journal of physical chemistry. A, 112(33), 7567-7576 (2008-07-26)
The contributions of terephthalic acid and Zn(2+)-coordinated water in N,N-diethylformamide (DEF) to the overall proton activity in the synthesis of MOF-5 (Zn4O(BDC)3, BDC = 1,4-benzenedicarboxylate) were quantitatively determined by combined electrochemical and UV-vis spectroscopic measurements. Structural transformations of zinc carboxylate-based

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service