Skip to Content
Merck
All Photos(1)

Documents

748897

Sigma-Aldrich

2-(1H-1,2,4-Triazol-3-yl)pyridine

97%

Synonym(s):

2-(2H-[1,2,4]Triazol-3-yl)-pyridine, 3-(2-Pyridyl)-1H-1,2,4-triazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6N4
CAS Number:
Molecular Weight:
146.15
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

powder or crystals

mp

157-162 °C

SMILES string

c1ccc(nc1)-c2ncn[nH]2

InChI

1S/C7H6N4/c1-2-4-8-6(3-1)7-9-5-10-11-7/h1-5H,(H,9,10,11)

InChI key

PVMLJDLTWMGZAH-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liang, B.; et al.
Journal of Materials Chemistry, 16, 1281-1286 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service