Skip to Content
Merck
All Photos(2)

Key Documents

748471

Sigma-Aldrich

2-Amino-3-bromobenzoic acid

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€43.10

€43.10


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€43.10

About This Item

Empirical Formula (Hill Notation):
C7H6BrNO2
CAS Number:
Molecular Weight:
216.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€43.10


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

174-178 °C

functional group

bromo
carboxylic acid

SMILES string

Nc1c(Br)cccc1C(O)=O

InChI

1S/C7H6BrNO2/c8-5-3-1-2-4(6(5)9)7(10)11/h1-3H,9H2,(H,10,11)

InChI key

SRIZNTFPBWRGPB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

322.7 °F

Flash Point(C)

161.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service