Skip to Content
Merck
All Photos(1)

Key Documents

748323

Sigma-Aldrich

3-Bromophenanthrene

96%

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€90.10

€90.10


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€90.10

About This Item

Empirical Formula (Hill Notation):
C14H9Br
CAS Number:
Molecular Weight:
257.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€90.10


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

96%

form

solid

mp

81-86 °C

functional group

bromo

SMILES string

Brc1ccc2ccc3ccccc3c2c1

InChI

1S/C14H9Br/c15-12-8-7-11-6-5-10-3-1-2-4-13(10)14(11)9-12/h1-9H

InChI key

BNGNNFQSUWVWCW-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rong Jin et al.
Journal of chromatography. A, 1509, 114-122 (2017-06-18)
Isotopic dilution gas chromatography combined with high resolution mass spectrometry (GC/HRMS) has overwhelming advantages with respect to the accuracy of congener-specific ultratrace analysis of complex persistent organic pollutants (POPs) in environmental matrices. However, an isotopic dilution GC/HRMS method for analysis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service