747785
[Ru(bpm)3][Cl]2
Synonym(s):
Dichlorotris(2,2′-bipyrimidine-N1,N1′)ruthenium(II), Tris(2,2′-bipyrimide)ruthenium(II) dichloride
About This Item
Recommended Products
form
powder
Quality Level
reaction suitability
core: ruthenium
reagent type: catalyst
reaction type: Photocatalysis
impurities
≤15 wt. % ethylene glycol
mp
>300 °C
SMILES string
C1(C2=NC=CC=N2)=NC=CC=N1.C3(C4=NC=CC=N4)=NC=CC=N3.C5(C6=NC=CC=N6)=NC=CC=N5.[Cl-].[Cl-].[Ru+2]
InChI
1S/3C8H6N4.2ClH.Ru/c3*1-3-9-7(10-4-1)8-11-5-2-6-12-8;;;/h3*1-6H;2*1H;/q;;;;;+2/p-2
InChI key
DVHNMJRXSLLELA-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
General description
Application
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
STOT RE 2 Oral
Target Organs
Kidney
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service