Skip to Content
Merck
All Photos(1)

Documents

747513

Sigma-Aldrich

N-Methyl-13C,d3-nicotinamide

98 atom % D, 99 atom % 13C, 98% (CP)

Synonym(s):

N-methyl-13C, d3-Vitamin B3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC6D3H5N2O
Molecular Weight:
140.16
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

99 atom % 13C
98 atom % D

Assay

98% (CP)

form

solid

technique(s)

mass spectrometry (MS): suitable

mass shift

M+4

storage temp.

2-8°C

SMILES string

O=C(N[13C]([2H])([2H])[2H])C1=CN=CC=C1

InChI

1S/C7H8N2O/c1-8-7(10)6-3-2-4-9-5-6/h2-5H,1H3,(H,8,10)/i1+1D3

InChI key

ZYVXHFWBYUDDBM-KQORAOOSSA-N

General description

N-Methyl nicotinamide is a metabolite of nicotinic acid which is found in human plasma and is a potent anti-inflammatory agent.
N-Methyl-13C,d3-nicotinamide is an isotope of vitamin B3 wherein all the three protons and the carbon atom of the N-methyl group are replaced by deuterium and 13C6 isotope, respectively.

Application

N-Methyl-13C,d3-nicotinamide can be used as a stable isotope internal standard for data interpretation in specific biological samples.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Topical application of 1-methylnicotinamide in the treatment of rosacea: a pilot study
Wozniacka A, et al.
Clinical and Experimental Dermatology, 30(6), 632-635 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service