Skip to Content
Merck
All Photos(1)

Documents

745766

Sigma-Aldrich

11-Mercaptoundecanamide

97%

Synonym(s):

10-Amido-1-decanethiol, Undecanamide, 11-mercapto-

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H23NOS
CAS Number:
Molecular Weight:
217.37
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

88-94 °C

storage temp.

2-8°C

SMILES string

NC(=O)CCCCCCCCCCS

InChI

1S/C11H23NOS/c12-11(13)9-7-5-3-1-2-4-6-8-10-14/h14H,1-10H2,(H2,12,13)

InChI key

LBHLMQVBCVDGID-UHFFFAOYSA-N

Application

“Wave function engineering” through charge transport by tunneling across self-assembled monolayers (SAMs) for molecular electronics.
Amido derivative SAMs have been used to control the protein absorption in biological applications.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thuo;Martin M.;
Journal of the American Chemical Society, 134(26), 10876-10884 (2101)
Ananthakrishnan Sethuraman et al.
Langmuir : the ACS journal of surfaces and colloids, 20(18), 7779-7788 (2004-08-25)
Besides significantly broadening the scope of available data on adhesion of proteins on solid substrates, we demonstrate for the first time that all seven proteins (tested here) behave similarly with respect to adhesion exhibiting a step increase in adhesion as
Sigal;George B.;
Journal of the American Chemical Society, 120(14), 3464-3473 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service