Skip to Content
Merck
All Photos(1)

Key Documents

744891

Sigma-Aldrich

Disodium ethyl/butyl phosphonate Silica

≥45 μm particle size (95%)

Synonym(s):

PhosphonicS PO1

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
12352200
NACRES:
NA.22

form

powder

extent of labeling

0.7-1.3 mmol/g loading

particle size

≥45 μm (95%)

General description

Disodium ethyl/butyl phosphonate Silica is an alkyl phosphonate functionalized silica with metal scavenging ability.

Application

Silica Supported Metal Scavenger:
Mg(II), Al(III), Ca(II), VO(II), Cr(III), Mn(II), Fe(III), Co(II), Ni(II), Cu(II), Zn(II), Sr(II), Ag(I), Cd(II), Sn(II), Sn(IV), La(III), Ce(III), Ce(IV), Hg(II), Pb(II), UO2(II)

Analysis Note

sodium: detected

Legal Information

Sold in collaboration with PhosphonicS Ltd. for research purposes only.
PhosphonicS is a trademark of PhosphonicS Ltd.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Muta. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service