Skip to Content
Merck
All Photos(2)

Documents

741469

Sigma-Aldrich

4-(1HTetrazol-5-yl)phenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7BN4O2
CAS Number:
Molecular Weight:
189.97
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

mp

174-179 °C (Decomposition)

storage temp.

2-8°C

SMILES string

OB(O)c1ccc(cc1)-c2nnn[nH]2

InChI

1S/C7H7BN4O2/c13-8(14)6-3-1-5(2-4-6)7-9-11-12-10-7/h1-4,13-14H,(H,9,10,11,12)

InChI key

DXUPJOQUAAVAGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Product is used as a reactant for palladium-catalyzed carbon-carbon bond formation (Suzuki-Miyuara reaction).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deborah S Mortensen et al.
Bioorganic & medicinal chemistry letters, 21(22), 6793-6799 (2011-10-08)
We report here the discovery of a novel series of selective mTOR kinase inhibitors. A series of imidazo[4,5-b]pyrazin-2-ones, represented by screening hit 1, was developed into lead compounds with excellent mTOR potency and exquisite kinase selectivity. Potent compounds from this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service