740578
1α,25-Dihydroxyvitamin D3 (6,19,19-d3) solution
100 μg/mL in ethanol, 97 atom % D, 96% (CP)
Synonym(s):
1α,25-Dihydroxycholecalciferol (6,19,19-d3) solution, Calcitriol (6,19,19-d3) solution, Deuterated 1-α25-Dihydroxyvitamin D3 solution
About This Item
Recommended Products
isotopic purity
97 atom % D
Quality Level
Assay
96% (CP)
form
solution
concentration
100 μg/mL in ethanol
technique(s)
mass spectrometry (MS): suitable
color
colorless
application(s)
detection
mass shift
M+3
storage temp.
−20°C
SMILES string
[2H]/C([2H])=C1[C@@H](O)C[C@H](O)CC\1=C([2H])\C=C2/CCC[C@]3(C)[C@H](CC[C@@H]23)[C@H](C)CCCC(C)(C)O
InChI
1S/C27H44O3/c1-18(8-6-14-26(3,4)30)23-12-13-24-20(9-7-15-27(23,24)5)10-11-21-16-22(28)17-25(29)19(21)2/h10-11,18,22-25,28-30H,2,6-9,12-17H2,1,3-5H3/b20-10+,21-11-/t18-,22-,23-,24+,25+,27-/m1/s1/i2D2,11D
InChI key
GMRQFYUYWCNGIN-POLIPXRRSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
1α,25-Dihydroxyvitamin D3 (6,19,19-d3) is a deuterated vitamin D3 wherein C-6 and C-19 protons are replaced by deuterium.
Application
Packaging
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
57.2 °F - closed cup
Flash Point(C)
14.0 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service