Skip to Content
Merck
All Photos(1)

Documents

740284

Sigma-Aldrich

Vitamin D3(6,19,19-d3) solution

100 μg/mL in ethanol, 97 atom % D, 97% (CP)

Synonym(s):

Calciol (6,19,19-d3) solution, Cholecalciferol (6,19,19-d3) solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27D3H41O
CAS Number:
Molecular Weight:
387.66
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.24

isotopic purity

97 atom % D

Assay

97% (CP)

form

solution

concentration

100 μg/mL in ethanol

technique(s)

mass spectrometry (MS): suitable

color

colorless

application(s)

detection

mass shift

M+3

storage temp.

−20°C

SMILES string

[2H]/C([2H])=C1CC[C@H](O)CC\1=C([2H])\C=C2/CCC[C@]3(C)[C@H](CC[C@@H]23)[C@H](C)CCCC(C)C

InChI

1S/C27H44O/c1-19(2)8-6-9-21(4)25-15-16-26-22(10-7-17-27(25,26)5)12-13-23-18-24(28)14-11-20(23)3/h12-13,19,21,24-26,28H,3,6-11,14-18H2,1-2,4-5H3/b22-12+,23-13-/t21-,24+,25-,26+,27-/m1/s1/i3D2,13D

InChI key

QYSXJUFSXHHAJI-GLSUUORTSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Deuterated vitamin D3(6,19,19-d3) solution is also known as cholecalciferol (6, 19, 19-d3). It can be used as a stable isotope internal standard for the quantification of vitamin D in biological samples by a liquid chromatography-tandem mass spectrometry (LC-MS/MS).
This product may be used as an analytical standard.

Packaging

1 mL in ampule

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potential intake of vitamins ?A? and ?D? through branded intravenous lipid emulsions: Liquid Chromatography-Tandem Mass Spectrometry Analysis
Forchielli ML, et al.
Clinical Nutrition (Edinburgh, Scotland), 36(2), 530-536 (2017)
Mechanism of error caused by isotope-labeled internal standard: accurate method for simultaneous measurement of vitamin D and pre-vitamin D by liquid chromatography/tandem mass spectrometry
Huang M, et al.
Rapid Communications in Mass Spectrometry, 28(19), 2101-2110 (2014)
Linda Ahonen et al.
Journal of chromatography. A, 1364, 214-222 (2014-09-11)
We have developed an ultra-high-performance liquid chromatography-atmospheric pressure photoionization-tandem mass spectrometric (UHPLC-APPI-MS/MS) method for the simultaneous quantitative analyses of several oxysterols and vitamin D metabolites in mouse brain and cell line samples. An UHPLC-APPI-high resolution mass spectrometric (UHPLC-APPI-HRMS) method that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service