Skip to Content
Merck
All Photos(1)

Documents

738425

Sigma-Aldrich

Potassium 3-methoxyphenyltrifluoroborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3COC6H4BF3 ·K
CAS Number:
Molecular Weight:
214.03
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

178-183 °C

SMILES string

[K+].COc1cccc(c1)[B-](F)(F)F

InChI

1S/C7H7BF3O.K/c1-12-7-4-2-3-6(5-7)8(9,10)11;/h2-5H,1H3;/q-1;+1

InChI key

GKDCWEPJOSDGHY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service