Skip to Content
Merck
All Photos(2)

Key Documents

737720

Sigma-Aldrich

Z-Gly-Bt

97%

Synonym(s):

N-Z-(1-Benzotriazolylcarbonyl)methylamine, Benzyl 2-(1H-benzo[d][1,2,3]triazol-1-yl)-2-oxoethylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14N4O3
CAS Number:
Molecular Weight:
310.31
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

mp

110-117 °C

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

O=C(NCC(=O)n1nnc2ccccc12)OCc3ccccc3

InChI

1S/C16H14N4O3/c21-15(20-14-9-5-4-8-13(14)18-19-20)10-17-16(22)23-11-12-6-2-1-3-7-12/h1-9H,10-11H2,(H,17,22)

InChI key

GXIYFGDLDAFXEF-UHFFFAOYSA-N

Application

Benzotriazole amino acids, or aminoacylbenzotriazolides, are versatile reagents for synthesizing peptides as well as their mimetics and conjugates. Benzotriazole amino acids have been used for the preparation of diverse derivatives including:

  • Polypeptidal benzotriazolides
  • Peptidomimetics, such as aminoxypeptides, depsipeptides and heterocyclic peptidomimetics
  • Tagged peptides and peptidomimetics, particularly those with fluorescent labels
  • N, O, S, and C linked peptide conjugates

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katritzky, A. R.; Narindoshvili, T.; Angrish, P.
Synthesis, 2013-2022 (2008)
Hansen, F. K.; Beagle, L. K.; Todadze, E.; Katritzky, A. R.
Heterocycles, 515-526 (2012)
Abdelmajeid, A.; Tala, S. R.; Amine, M. S.; Katritzky, A. R.
Synthesis, 2995-3005 (2011)
Katritzky, A. R.; Angrish, P.; Todadze, E.
Synlett, 2392-2411 (2009)
Ilker Avan et al.
The Journal of organic chemistry, 76(12), 4884-4893 (2011-04-02)
Reactions of O-Pg(α-hydroxyacyl)benzotriazoles with (a) unprotected α-hydroxycarboxylic acids, (b) amino acids, and (c) amines afforded, respectively, chirally pure (a) oligoesters, (b) depsidipeptides, and (c) amide conjugates (yields 52-94%). N-Pg(α-Aminoacyl)benzotriazoles reacted with α-hydroxycarboxylic acids to yield depsidipeptides (47-87%). N-Pg(depsidipeptidoyl)benzotriazoles, obtained from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service