Skip to Content
Merck
All Photos(1)

Key Documents

736988

Sigma-Aldrich

1-Amino-1-(hydroxyethyl)cyclohexane

97%

Synonym(s):

1-Amino-1-cyclohexanemethanol

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€91.50

€91.50


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€91.50

About This Item

Empirical Formula (Hill Notation):
C7H15NO
CAS Number:
Molecular Weight:
129.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€91.50


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

34-38 °C

functional group

hydroxyl

SMILES string

NC1(CO)CCCCC1

InChI

1S/C7H15NO/c8-7(6-9)4-2-1-3-5-7/h9H,1-6,8H2

InChI key

VDPLLINNMXFNQX-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service