Skip to Content
Merck
All Photos(1)

Key Documents

736945

Sigma-Aldrich

5-Methyl-3,4-hexadien-1-ol

95%

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€71.90

€71.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€71.90

About This Item

Empirical Formula (Hill Notation):
C7H12O
CAS Number:
Molecular Weight:
112.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€71.90


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.477

density

0.863 g/mL at 25 °C

functional group

hydroxyl

storage temp.

2-8°C

SMILES string

CC(C)=C=CCCO

InChI

1S/C7H12O/c1-7(2)5-3-4-6-8/h3,8H,4,6H2,1-2H3

InChI key

JPVXYBRKKQSXLO-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids not in Storage Class 3

WGK

WGK 3

Flash Point(F)

150.1 °F

Flash Point(C)

65.6 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service