Skip to Content
Merck
All Photos(1)

Key Documents

736503

Sigma-Aldrich

Silver, dispersion

nanoparticle, 50-60 wt. % in tetradecane, spec. resistivity ~ 2.7 μΩ-cm, for printing on polyimide films

Synonym(s):

Silverjet DGH-55LT-25C, silver ink

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

description

surface tension 27 - 31 dyn/cm

form

dispersion
nanoparticle

concentration

50-60 wt. % in tetradecane

refractive index

n20/D 1.333

particle size

≤10 nm

viscosity

7-14 cP

density

1.5-1.8 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silver is a good conductor and is resistant to oxidation. It is used in printed conductors and printable inks (based on silver nanoparticles). Nanosilver is used as an antibacterial material in filters for air and water purification, in textiles, and for wound treatment. Other large scale applications of nanosilver include metallization of solar cells, thin film electronics, sensors, and catalysts.

Curing Temperature: 250 °C

Recommended substrates: Polyimide films

Recommended washing solvent: Non-Polar Solvents: Xylene, Hexane, Octane etc. (Hydrocarbon)

Legal Information

Product of ANP, Inc.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

219.9 °F

Flash Point(C)

104.4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Articles

Inkjet printing is one of the key enabling technologies of printed electronics. Inkjet printing technology classification, aspects of materials (inks, substrates) and respective pre-and post-processing steps are discussed.

Functional materials for printed electronics applications enable flexible displays, RFID tags, and biomedical sensors.

The ability to pattern conductive electrodes is technologically relevant for several applications, including photovolatics, displays, sensors, and biomedical devices.

Small molecular weight organic semiconductors are promising for flexible transistor applications in next-gen soft electronics.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service