Skip to Content
Merck
All Photos(1)

Key Documents

736481

Sigma-Aldrich

Silver, dispersion

nanoparticle, ≤50 nm particle size, 30-35 wt. % in triethylene glycol monoethyl ether, spec. resistivity ~ 2 μΩ-cm, for printing on ITO and glass

Synonym(s):

Silverjet DGP-45HTG, silver ink

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

description

surface tension 30 - 50 dyn/cm

form

nanoparticle

concentration

30-35 wt. % in triethylene glycol monoethyl ether

refractive index

n20/D 1.333

particle size

≤50 nm

viscosity

10-18 cP

density

1.45 g/mL±0.05 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silver is a good conductor and resistant to oxidation. It is used in printed conductors and printable inks (based on silver nanoparticles). Nanosilver is used as an antibacterial material in filters for air and water purification, in textiles, and for wound treatment. Other large scale applications of nanosilver include metallization of solar cells, thin film electronics, sensors, and catalysts.

Curing Temperature: Above 400 °C

Recommended substrates: ITO, Glass, Etc.

Recommended washing solvent: Polar Solvents: Ethyl Alcohol, IPA etc.

Legal Information

Product of ANP, Inc.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Inkjet printing is one of the key enabling technologies of printed electronics. Inkjet printing technology classification, aspects of materials (inks, substrates) and respective pre-and post-processing steps are discussed.

Functional materials for printed electronics applications enable flexible displays, RFID tags, and biomedical sensors.

The ability to pattern conductive electrodes is technologically relevant for several applications, including photovolatics, displays, sensors, and biomedical devices.

Progress in solution-processed functional materials leads to thin-film optoelectronic devices for industrial and consumer electronics.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service