736333
Poly(ethylene glycol) methyl ether 2-bromoisobutyrate
average Mn 5,000
Synonym(s):
Polyethylene glycol, ATRP initiator, PEG methyl ether bromoisobutyrate, mPEG bromoisobutyrate
About This Item
Recommended Products
form
solid
mol wt
average Mn 5,000
transition temp
Tm 56-60 °C
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
ATRP is a successful method for precise polymer synthesis with controlled molecular weights and high chain end functionalities.
Over the past two decades, the rapid advance of controlled living polymerization (CLP) techniques.
Protocols
Polymerization via ATRP procedures demonstrated by Prof. Dave Haddleton's research group at the University of Warwick.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service