Skip to Content
Merck
All Photos(1)

Key Documents

736155

Sigma-Aldrich

Potassium oleate-d33

98 atom % D

Synonym(s):

Oleic acid-d33 potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3(CD2)7CD=CD(CD2)7COOK
Molecular Weight:
353.76
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

98 atom % D

form

powder

mass shift

M+33

storage temp.

2-8°C

SMILES string

[K+].[2H]\C(=C(/[2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([O-])=O

InChI

1S/C18H34O2.K/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h9-10H,2-8,11-17H2,1H3,(H,19,20);/q;+1/p-1/b10-9-;/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D,10D,11D2,12D2,13D2,14D2,15D2,16D2,17D2;

InChI key

MLICVSDCCDDWMD-IEKXYDGZSA-M

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

284.0 °F - closed cup

Flash Point(C)

140.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service