Skip to Content
Merck
All Photos(1)

Key Documents

733865

Sigma-Aldrich

Aldicarb-(N-methyl-13C,d3 carbamoyl-13C)

≥98 atom %, ≥98% (CP)

Synonym(s):

(1E)-2-Methyl-2-(methylthio)-O-[(methyl-13C-d3-amino)carbonyl-13C]oxime propanal

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€404.00
5 MG
€884.00
10 MG
€1,630.00
25 MG
€3,740.00

€404.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 MG
€404.00
5 MG
€884.00
10 MG
€1,630.00
25 MG
€3,740.00

About This Item

Empirical Formula (Hill Notation):
13C2C5D3H11N2O2S
CAS Number:
Molecular Weight:
195.27
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

€404.00


Please contact Customer Service for Availability

Request more information

isotopic purity

≥98 atom %

Assay

≥98% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mass shift

M+5

storage temp.

−20°C
−20°C

SMILES string

[2H][13C]([2H])([2H])N[13C](=O)O\N=C\C(C)(C)SC

InChI

1S/C7H14N2O2S/c1-7(2,12-4)5-9-11-6(10)8-3/h5H,1-4H3,(H,8,10)/b9-5+/i3+1D3,6+1

InChI key

QGLZXHRNAYXIBU-FONWFMSDSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service