Skip to Content
Merck
All Photos(1)

Key Documents

732907

Sigma-Aldrich

3,7-Dibromochromone

Synonym(s):

3,7-Dibromo-4H-benzopyran-4-one, 3,7-Dibromo-4H-chromen-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H4Br2O2
CAS Number:
Molecular Weight:
303.93
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

140-146 °C

SMILES string

Brc1ccc2C(=O)C(Br)=COc2c1

InChI

1S/C9H4Br2O2/c10-5-1-2-6-8(3-5)13-4-7(11)9(6)12/h1-4H

InChI key

VRJRKKZNBPQORR-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service