Skip to Content
Merck
All Photos(2)

Key Documents

731706

Sigma-Aldrich

3,3′,5,5′-Tetracarboxydiphenylmethane

greener alternative

≥95% (HPLC)

Synonym(s):

Linker for PCN-12 MOF

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€144.00

€144.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€144.00

About This Item

Empirical Formula (Hill Notation):
C17H12O8
CAS Number:
Molecular Weight:
344.27
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

€144.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

solid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

324-331 °C

greener alternative category

SMILES string

OC(=O)c1cc(Cc2cc(cc(c2)C(O)=O)C(O)=O)cc(c1)C(O)=O

InChI

1S/C17H12O8/c18-14(19)10-2-8(3-11(6-10)15(20)21)1-9-4-12(16(22)23)7-13(5-9)17(24)25/h2-7H,1H2,(H,18,19)(H,20,21)(H,22,23)(H,24,25)

InChI key

RAESDWWKTFZWJA-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enhancing H2 uptake by "close-packing" alignment of open copper sites in metal-organic frameworks.
Xi-Sen Wang et al.
Angewandte Chemie (International ed. in English), 47(38), 7263-7266 (2008-08-21)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service