Skip to Content
Merck
All Photos(1)

Key Documents

731668

Sigma-Aldrich

Estriol-2,3,4-13C3

99 atom % 13C, 97% (CP)

Synonym(s):

16-α-Hydroxyestradiol-2,3,4-13C3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€765.00
250 μG
€1,530.00
500 μG
€2,760.00
1000 μG
€4,740.00

€765.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
100 μG
€765.00
250 μG
€1,530.00
500 μG
€2,760.00
1000 μG
€4,740.00

About This Item

Empirical Formula (Hill Notation):
13C3C15H24O3
CAS Number:
Molecular Weight:
291.36
MDL number:
UNSPSC Code:
12142200
PubChem Substance ID:
NACRES:
NA.24

€765.00


Please contact Customer Service for Availability

Request more information

isotopic purity

99 atom % 13C

Quality Level

Assay

97% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mp

280-282 °C (lit.)

mass shift

M+3

storage temp.

−20°C

SMILES string

C[C@]12CC[C@H]3[C@@H](CCc4[13cH][13c](O)[13cH]cc34)[C@@H]1C[C@@H](O)[C@@H]2O

InChI

1S/C18H24O3/c1-18-7-6-13-12-5-3-11(19)8-10(12)2-4-14(13)15(18)9-16(20)17(18)21/h3,5,8,13-17,19-21H,2,4,6-7,9H2,1H3/t13-,14-,15+,16-,17+,18+/m1/s1/i3+1,8+1,11+1

InChI key

PROQIPRRNZUXQM-FDFADMNFSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service