Skip to Content
Merck
All Photos(1)

Key Documents

727229

Sigma-Aldrich

(R)-3,4-Dimethoxy-α-methylbenzylamine

ChiPros®, produced by BASF, 99%

Synonym(s):

(R)-1-(3,4-Dimethoxyphenyl)ethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15NO2
CAS Number:
Molecular Weight:
181.23
Beilstein:
8616003
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥98.5% (GC)
99%

form

liquid

optical purity

enantiomeric excess: ≥98.5%

SMILES string

COc1ccc(cc1OC)[C@@H](C)N

InChI

1S/C10H15NO2/c1-7(11)8-4-5-9(12-2)10(6-8)13-3/h4-7H,11H2,1-3H3/t7-/m1/s1

InChI key

OEPFPKVWOOSTBV-SSDOTTSWSA-N

Legal Information

ChiPros is a registered trademark of BASF SE

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service