Skip to Content
Merck
All Photos(1)

Key Documents

725897

Sigma-Aldrich

2-(Chloromethyl)benzoyl chloride

≥96.0% (GC)

Synonym(s):

α-Chloro-o-toluoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6Cl2O
CAS Number:
Molecular Weight:
189.04
Beilstein:
1940781
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96.0% (GC)
96.0-104.0% (T)

form

liquid

functional group

acyl chloride
chloro

SMILES string

ClCc1ccccc1C(Cl)=O

InChI

1S/C8H6Cl2O/c9-5-6-3-1-2-4-7(6)8(10)11/h1-4H,5H2

InChI key

TXZFBHYDQGYOIT-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jae Young Jeong et al.
Acta biomaterialia, 53, 414-426 (2017-02-22)
(3-Aminomethylphenyl)boronic acid (AMPB)-installed hyaluronic acid-ceramide (HACE)-based nanoparticles (NPs), including manassantin B (MB), were fabricated for tumor-targeted delivery. The amine group of AMPB was conjugated to the carboxylic acid group of hyaluronic acid (HA) via amide bond formation, and synthesis was
Joo-Eun Kim et al.
International journal of nanomedicine, 12, 645-658 (2017-02-09)
Paclitaxel-loaded hyaluronan solid nanoemulsions (PTX-HSNs) were successfully fabricated for the delivery of PTX to improve ovarian cancer treatment via active tumor targeting. PTX-HSNs were fabricated using high-pressure homogenization with a microfluidizer and were lyophilized with d-mannitol. Hyaluronan was coated on
Song Yi Lee et al.
ACS applied materials & interfaces, 9(27), 22308-22320 (2017-06-18)
An amphiphilic hyaluronic acid-ceramide-dopamine (HACE-d) conjugate was prepared, and HACE-d-based nanoparticles (NPs) including phloretin (as an inhibitor of glucose transporter (GLUT1)) were fabricated. Mussel-inspired property of d was introduced to HACE NPs, and it may improve tumor targetability and penetration
Joo-Eun Kim et al.
Colloids and surfaces. B, Biointerfaces, 150, 362-372 (2016-11-09)
The purpose of this study was to develop hyaluronan-coated nanoemulsions (HNEs) with high solubilizing capacity and tumor cell targeting capability for the poorly soluble paclitaxel. The HNEs were composed of dl-a-tocopheryl acetate, soybean oil, polysorbate 80, and ferric chloride and
Joo-Eun Kim et al.
Biomolecules & therapeutics, 25(4), 411-416 (2017-02-17)
Paclitaxel (PTX) is a effectively chemotherapeutic agent which is extensively able to treat the non-small cell lung, pancreatic, breast and other cancers. But it is a practically insoluble drug with water solubility less than 1 µg/mL, which restricts its therapeutic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service