Skip to Content
Merck
All Photos(1)

Key Documents

725382

Sigma-Aldrich

1,6-Hexanedithiol

99.5%

Synonym(s):

1,6-Dimercaptohexane, DMH, Hexamethylene dimercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HS(CH2)6SH
CAS Number:
Molecular Weight:
150.31
Beilstein:
1732507
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

~1 mmHg ( 20 °C)

Assay

99.5%

form

liquid

refractive index

n20/D 1.511 (lit.)

bp

118-119 °C/15 mmHg (lit.)

mp

−21 °C (lit.)

density

0.983 g/mL at 25 °C (lit.)

SMILES string

SCCCCCCS

InChI

1S/C6H14S2/c7-5-3-1-2-4-6-8/h7-8H,1-6H2

InChI key

SRZXCOWFGPICGA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,6-Hexanedithiol (HDT) is a long chained bi-functional alkanethiol with 6 carbon atoms structured in a zig-zag pattern, which forms a tetrahedral configuration. It forms a self-assembled monolayer (SAM) on a variety of substrates and immobilizes the surface atoms.

Application

HDT forms a SAM on Au, Pt, and Ag substrates, which can be used as coated electrode materials for electrochemical and biological detections. These electrodes form a sensing platform that can be used in biomedical and biosensing applications.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nucleic acid-functionalized Pt nanoparticles: catalytic labels for the amplified electrochemical detection of biomolecules
Polsky R, et al.
Analytical Chemistry, 78(7), 2268-2271 (2006)
Combined TPD and XPS Study of Ligation and Decomposition of 1, 6-Hexanedithiol on Size-Selected Copper Clusters Supported on HOPG.
Wang L, et al.
The Journal of Physical Chemistry C, 122(4), 2173-2183 (2018)
Length-dependent transport in molecular junctions based on SAMs of alkanethiols and alkanedithiols: effect of metal work function and applied bias on tunneling efficiency and contact resistance
Engelkes VB, et al.
Journal of the American Chemical Society, 126(43), 14287-14296 (2004)
Covalent attachment of glucose oxidase to an Au electrode modified with gold nanoparticles for use as glucose biosensor
Zhang S, et al.
Bioelectrochemistry, 67(1), 15-22 (2005)
1, 6-hexanedithiol monolayers on Au (111): A multitechnique structural study.
Leung TYB, et al.
Langmuir, 16(2), 549-561 (2000)

Articles

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service